Farnesol (mixture of isomers) 96%

Synonyms: Farnesol2,6,10-Dodecatrien-1-ol, 3,7,11-trimethyl-, (2Z,6Z)-3,7,11-Trimethyldodeca-2,6,10-trien-1-olTRANS,TRANS-3,7,11-TRIMETHYL-2,6,10-DODECATRIEN-1-OL

AC11912-0250 AC11912-1000
200012-510EA 39.4 USD
200012-510 200012-512
Farnesol (mixture of isomers) 96%
Farnesol (mixture of isomers)
Formula: C₁₅H₂₆O
MW: 222.37 g/mol
Boiling Pt: 149 °C (4 mmHg)
Density: 0.8865 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00002918
CAS Number: 4602-84-0
EINECS: 225-004-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4920 (20 °C, 589 nm)
Density (20/4°C) 0.884 to 0.89 kg/l
GC ≥95 % (sum of geometric isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR