Ethyl piperazine-1-carboxylate 99%
|
Warning
|
Synonyms:
1-Ethoxycarbonyl piperazine
, Ethyl N-piperazinecarboxylate
AC11887-1000
AC11887-0250
200012-430EA
110
USD
200012-430
200012-428
Ethyl piperazine-1-carboxylate 99%
Ethyl piperazine-1-carboxylate
Formula:
C₇H₁₄N₂O₂ MW: 158.2 g/mol Boiling Pt: 273 °C (760 mmHg) Density: 1.08 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005964 CAS Number: 120-43-4 EINECS: 204-395-2 |
Specification Test Results
Appearance | Clear colorless to light yellow Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4760 to 1.4790 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...