Ethyl picolinate 99%

Synonyms: Ethyl 2-pyridinecarboxylateEthyl 2-picolinate

AC11886-0250 AC11886-1000
200012-434EA 59.1 USD
200012-434 200012-436
Ethyl picolinate 99%
Ethyl picolinate
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 2 °C
Density: 1.11 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00006292
CAS Number: 2524-52-9
EINECS: 219-758-0

Order Now

Specification Test Results

Appearance Clear yellow to pink Liquid
Infrared spectrum Conforms
Refractive index 1.5090 to 1.5110 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR