Ethyl phenylcyanoacetate 97%
|
Warning
|
Synonyms:
Ethyl 2-cyano-2-phenylacetate
AC11883-0250
AC11883-0050
200043-520EA
175
USD
200043-520
200043-518
Ethyl phenylcyanoacetate 97%
Ethyl phenylcyanoacetate
Formula:
C₁₁H₁₁NO₂ MW: 189.21 Density: 1.09 g/cm³ Storage Temperature: Ambient |
EINECS:
224-921-4 UN: 2810 ADR: 6.1,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5033 to 1.5053 (20 °C, 589 nm) |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...