Ethyl oxalyl chloride 98%

Danger

Synonyms: Ethyl oxalyl monochlorideChloroglyoxylic Acid Ethyl EsterEthyl ChloroglyoxylateOxalic Acid Monoethyl Ester ChlorideEthyl chloroformylformateEthyl Chlorooxoacetate

AC11878-1000 AC11878-5000 AC11878-0250
200006-502EA 65.9 USD
200006-502 200006-500 103530-644
Ethyl oxalyl chloride 98%
Ethyl oxalyl chloride
Formula: C₄H₅ClO₃
MW: 136.53 g/mol
Boiling Pt: 135 °C (760 mmHg)
Density: 1.22 g/cm³
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00000706
CAS Number: 4755-77-5
EINECS: 225-285-0
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4160 to 1.4180 (20 °C, 589 nm)
GC ≥97.5 %
Color scale ≤200 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR