Ethyl mercaptoacetate 99%
|
Danger
|
Synonyms:
Ethyl thioglycolate
, Ethyl 2-mercaptoacetate
AC11867-1000
AC11867-0025
AC11867-5000
200006-450EA
41.1
USD
200006-450
103530-642
200019-044
Ethyl mercaptoacetate 99%
Ethyl mercaptoacetate
Formula:
C₄H₈O₂S MW: 120.17 g/mol Boiling Pt: 54 °C (12 mmHg) Melting Pt: –80 °C Density: 1.09 g/cm³ Flash Pt: 47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004874 CAS Number: 623-51-8 EINECS: 210-800-3 UN: 1228 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4560 to 1.4590 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...