Ethyl (ethoxymethylene)cyanoacetate 98%

Danger

Synonyms: Ethyl 2-Cyano-3-Ethoxyacrylate

AC11848-1000 AC11848-5000
200037-424EA 165 USD
200037-424 200012-458
Ethyl (ethoxymethylene)cyanoacetate 98%
Ethyl (ethoxymethylene)cyanoacetate
Formula: C₈H₁₁NO₃
MW: 169.18 g/mol
Boiling Pt: 190…191 °C (30 mmHg)
Melting Pt: 48…52 °C
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00009136
CAS Number: 94-05-3
EINECS: 202-299-5
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to yellow-brown Powder or crystals or flakes or crystalline solid
Infrared spectrum Conforms
Melting point 47 to 53 °C
GC ≥97.5 %
GC ≥99.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 passes test
Sulfur compounds (as S) ≤0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR