Ethyl-2-oxocyclohexanecarboxylate 95%

Warning

Synonyms: Ethyl 2-oxocyclohexanecarboxylate

AC11833-1000 AC11833-0050 AC11833-0250
103509-470EA 89.8 USD
103509-470 200006-448 103508-524
Ethyl-2-oxocyclohexanecarboxylate 95%
Ethyl 2-oxocyclohexanecarboxylate
Formula: C₉H₁₄O₃
MW: 170.21 g/mol
Boiling Pt: 106 °C (11 mmHg)
Density: 1.06 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00001631
CAS Number: 1655-07-8
EINECS: 216-731-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4775 to 1.4795 (20 °C, 589 nm)
GC ≥94 %
Molybdenum (Mo) ≤10 ppm
Heavy metals & Fe (as Pb) ≤0.001 %
Titrable free base ≤0.02 meq/g
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR