Ethyl carbazate 97%
|
Warning
|
Synonyms:
Ethyl hydrazinecarboxylate
AC11820-1000
AC11820-5000
AC11820-0250
103530-628EA
294
USD
103530-628
200019-112
200019-110
Ethyl carbazate 97%
Ethyl carbazate
Formula:
C₃H₈N₂O₂ MW: 104.11 g/mol Boiling Pt: 108…110 °C (22 mmHg) Melting Pt: 44…48 °C Flash Pt: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007595 CAS Number: 4114-31-2 EINECS: 223-903-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to almost white (May have a pink hue) Crystals and/or chunks or fused solid |
Infrared spectrum | Conforms |
Melting point | 42 to 48 °C |
GC | ≥96.0 % |
Chloride (Cl) | ≤20 ppm |
Sulfate (SO4) | ≤50 ppm |
Iron (Fe) | ≤20 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Insoluble matter | ≤0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...