Ethyl-4-bromocrotonate 75% tech.

Danger

Synonyms: Ethyl trans-4-bromo-2-butenoate(E)-Ethyl 4-bromobut-2-enoate

AC11812-1000 AC11812-0250 AC11812-0050
103530-624EA 216 USD
103530-624 200025-388 103530-622
Ethyl-4-bromocrotonate 75% tech.
Ethyl 4-bromocrotonate
Formula: C₆H₉BrO₂
MW: 193.05 g/mol
Boiling Pt: 94…95 °C (12 mmHg)
Density: 1.4 g/cm³
Flash Pt: 97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000247
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
GC ≥70 %
Thiocyanate (SCN) ≤200 ppm
Phosphate (PO4) ≤200 ppm
Sulfate (SO4) ≤500 ppm
Lead (Pb) ≤5 ppm
Sulfur (S) ≤50 ppm
Iron, total (as Fe) ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR