3-Ethoxy-2-cyclohexen-1-one 99%

AC11788-0100 AC11788-1000 AC11788-0250
200009-912EA 89.6 USD
200009-912 103509-374 200023-270
3-Ethoxy-2-cyclohexen-1-one 99%
3-Ethoxy-2-cyclohexen-1-one
Formula: C₈H₁₂O₂
MW: 140.18 g/mol
Boiling Pt: 76…78 °C (1 mmHg)
Density: 1.04 g/cm³
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001580
CAS Number: 5323-87-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5020 to 1.5060 (20 °C, 589 nm)
GC ≥98.5 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR