Emodin 95% tech.

Warning

Synonyms: 1,3,8-Trihydroxy-6-methylanthracene-9,10-dione1,3,8-Trihydroxy-6-methylanthraquinone

AC11775-1000 AC11775-5000
200031-182EA 115 USD
200031-182 200019-008
Emodin 95% tech.
Emodin

Appearance: Orange-red Powder

Formula: C₁₅H₁₀O₅
MW: 270.24 g/mol
Melting Pt: 253 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001207
CAS Number: 518-82-1

Order Now

Specification Test Results

Appearance Orange-red Powder
Infrared spectrum Conforms
HPLC ≥94 %
Substance darkened by H2SO4 Passes test
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤100 ppm
Ammonium (NH4) ≤20 ppm
Iron (Fe) ≤10 ppm
Sodium (Na) ≤200 ppm
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.01 %
Neutrality Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR