2,2'-Dithiodi(benzoic acid) 98%
|
Warning
|
Synonyms:
2,2'-Dithiodibenzoic acid
, 2,2'-Dithiosalicylic acid
AC11756-5000
AC11756-1000
200034-170EA
117
USD
200034-170
200028-098
2,2'-Dithiodi(benzoic acid) 98%
2,2'-Dithiodi(benzoic acid)
Formula:
C₁₄H₁₀O₄S₂ MW: 306.36 g/mol Melting Pt: 287…290 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002465 CAS Number: 119-80-2 EINECS: 204-352-8 |
Specification Test Results
Appearance | Brown Powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥97.5 % |
Chloride (Cl) | ≤5 ppm |
Nitrate (NO3) | ≤0.002 % |
Ammonium (NH4) | ≤0.002 % |
Calcium (Ca) | ≤0.02 % |
Iron (Fe) | ≤5 ppm |
Manganese (Mn) | ≤5 ppm |
Potassium (K) | ≤0.005 % |
Sodium (Na) | ≤0.005 % |
Strontium (Sr) | ≤0.005 % |
Heavy metals (ICP-OES) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...