2,3-Diphenyl-1-indenone 97%

AC11732-0100
200034-284EA 101.8 USD
200034-284
2,3-Diphenyl-1-indenone 97%
2,3-Diphenyl-1-indenone

Appearance: Red Crystalline powder

Formula: C₂₁H₁₄O
Melting Pt: 151…155 °C
Storage Temperature: Ambient
CAS Number: 1801-42-9

Order Now

Specification Test Results

Appearance Red Crystalline powder
Infrared spectrum Conforms
Melting point 151 to 155 °C
GC ≥96.0 %
Phosphate (PO4) ≤1 ppm
SO4 and SO3 (as SO4) ≤5 ppm
Arsenic (As) ≤0.05 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤1 ppm
Heavy metals (as Pb) ≤0.5 ppm
Fluorosilicic acid ≤0.01 % (H2SiF6)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR