Diphenyl chlorophosphate 98%
|
Danger
|
Synonyms:
Diphenyl phosphorochloridate
AC11724-1000
AC11724-5000
AC11724-0250
200012-142EA
61.3
USD
200012-142
103525-020
200012-140
Diphenyl chlorophosphate 98%
Diphenyl chlorophosphate
Formula:
C₁₂H₁₀ClO₃P Boiling Pt: 314…316 °C (272 mmHg) Density: 1.29 g/cm³ Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003030 CAS Number: 2524-64-3 EINECS: 219-759-6 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5485 to 1.5505 (20 °C, 589 nm) |
GC | ≥97.5 % |
Sulfur compounds (as S) | ≤0.005 % |
Water soluble titr. acid | ≤0.0003 meq/g |
Thiophene | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...