Galactitol 99+%

Synonyms: Dulcitol

AC11770-5000 AC11770-1000 AC11770-0250
103509-354EA 287 USD
103509-354 103509-352 200012-304
Galactitol 99+%
Galactitol
Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 275…280 °C (1 mmHg)
Melting Pt: 188…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00064288
CAS Number: 608-66-2
EINECS: 210-165-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 187 to 191 °C
Water ≤1.0 % (K.F.)
Solubility (3 % in water) Colorless
HPLC ≥99.0 % (ELSD)
Heavy metals (as Pb) ≤0.003 %
Insoluble matter ≤0.01 % (in dilute NH4OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR