N,O-Dimethylhydroxylamine hydrochloride 98%

Warning

AC11634-1000 AC11634-0250 AC11634-0050
200000-412EA 352 USD
200000-412 200007-218 200000-410
N,O-Dimethylhydroxylamine hydrochloride 98%
N,O-Dimethylhydroxylamine hydrochloride
Formula: C₂H₈ClNO
MW: 97.55 g/mol
Melting Pt: 112…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00012485
CAS Number: 6638-79-5
EINECS: 229-642-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 112 to 116 °C
Water ≤0.5 %
Solubility (10 % in water) Clear
Titration with HClO4 ≥97.5 % (on anhydrous substance)
Sulfate (SO4) ≤0.003 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR