5,6-Dimethylbenzotriazole monohydrate 99%

Warning

Synonyms: 5,6-Dimethyl-1,2,3-benzotriazole Hydrate5,6-Dimethyl-1H-benzotriazole hydrate5,6-Dimethyl-1H-benzotriazole monohydrate

AC11605-0250 AC11605-0050
200036-156EA 47.8 USD
200036-156 200048-136
5,6-Dimethylbenzotriazole monohydrate 99%
5,6-Dimethylbenzotriazole monohydrate
Formula: C₈H₉N₃·1H₂O
MW: 165.20 g/mol (147.18 anhy)
Melting Pt: 153…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00149344
CAS Number: 4184-79-6
EINECS: 224-058-3

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 156 °C
Water ≤11 % (K.F.)
Titration with TBAH ≥98.5 % (on dry substance)
UV at 263 nm A: ≤0.1
UV at 275 nm A: ≤0.05
UV at 330 nm A: ≤0.01
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.003 %
Substance darkened by H2SO4 passes test
Titrable acid ≤0.0009 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR