2,6-Dimethyl-p-benzoquinone 97%

Warning

Synonyms: 2,6-Dimethylcyclohexa-2,5-diene-1,4-dione

AC11604-0010
200009-078EA 30 USD
200009-078
2,6-Dimethyl-p-benzoquinone 97%
2,6-Dimethyl-p-benzoquinone
Formula: C₈H₈O₂
MW: 136.15 g/mol
Melting Pt: 69…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00001605
CAS Number: 527-61-7
EINECS: 208-420-8

Order Now

Specification Test Results

Appearance Yellow to brown Crystals or needles and/or chunks
Infrared spectrum Conforms
Melting point 69 to 74 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR