Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloropropyldimethylammonium chloride ≥98.5%

Supplier: ACROS ORGANICS MS

Warning
Synonyms: N-(2-Chloropropyl)-N,N-dimethylammonium chloride2-Dimethylaminoisopropyl chloride hydrochloride

AC11583-1000 AC11583-5000
200041-010EA 25.55 USD InStock
200041-010 200046-818
2-Chloropropyldimethylammonium chloride ≥98.5%
2-Chloropropyldimethylammonium chloride


Formula: C₅H₁₃Cl₂N
MW: 158.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012534
CAS Number: 4584-49-0
EINECS: 224-971-7
Order Now
Specification Test Results
Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
Titration with HClO4 ≥98.0 % (On dry substance)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty