2-Chloroethyldimethylammonium chloride 99%

Warning

Synonyms: N-(2-Chloroethyl)-N,N-Dimethylammonium Chloride2-Dimethylaminoethyl chloride hydrochloride

AC11582-0050 AC11582-5000 AC11582-1000 AC11582-0025
200028-708EA 20.9 USD
200028-708 200005-040 103530-566 200022-620
2-Chloroethyldimethylammonium chloride 99%
2-Chloroethyldimethylammonium chloride
Formula: C₄H₁₁Cl₂N
MW: 144.04 g/mol
Melting Pt: 203…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00012516
CAS Number: 4584-46-7
EINECS: 224-970-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to brown-yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 203 to 210 °C
Water ≤1.0 % (K.F.)
Titration with NaOH 98.5 to 101.5 % (On dry substance)
Chloride (Cl) 24.1 to 25.1 % (ionic chloride)
Chloride (Cl) 48.4 to 50.0 % (total chloride)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR