2-(Dimethylamino)ethanethiol hydrochloride 95%

Warning

AC11581-1000 AC11581-0250 AC11581-0050
103509-266EA 428 USD
103509-266 103509-264 200016-544
2-(Dimethylamino)ethanethiol hydrochloride 95%
2-(Dimethylamino)ethanethiol hydrochloride
Formula: C₄H₁₂ClNS
Melting Pt: 154…159 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012633
EINECS: 236-221-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Iodimetric ≥94 %
GC ≥96.0 %
Enantiomeric excess ≥98.0 %
Specific optical rotation -7° to -9° (20 °C, 589 nm) (c=1, chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR