4-(Dimethylamino)benzonitrile 95%

Warning

AC11578-0250 AC11578-0050
200047-616EA 36.8 USD
200047-616 200023-480
4-(Dimethylamino)benzonitrile 95%
4-(Dimethylamino)benzonitrile
Formula: C₉H₁₀N₂
Melting Pt: 70…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00001815
CAS Number: 1197-19-9
EINECS: 214-819-8
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light brown Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 76 °C
Titration with HClO4 ≥94 %
Stabilizer About 500 ppm 4-tert-Butylcatecholo
Stabilizer About 500 ppm Nitrocresol + Nitrophenol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR