Dimethyl-3-oxoglutarate 97%

Warning

Synonyms: Dimethyl 1,3-acetonedicarboxylateDimethyl acetone-1,3-dicarboxylate

AC11570-5000 AC11570-1000 AC11570-0250
200000-384EA 85.5 USD
200000-384 103509-260 200012-094
Dimethyl-3-oxoglutarate 97%
Dimethyl 3-oxoglutarate
Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Boiling Pt: 237…241 °C (760 mmHg)
Melting Pt: 16…17 °C
Density: 1.18 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008462
CAS Number: 1830-54-2
EINECS: 217-385-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4450 (20 °C, 589 nm)
GC ≥96.0 %
Visual transition interval pH 5.5 (orange-yellow)
Visual transition interval pH 6.0 (yellow)
Visual transition interval pH 6.5 (yellow-green)
Visual transition interval pH 7.0 (green)
Visual transition interval pH 7.5 (green, slightly blue)
Visual transition interval pH 8.0 (green-blue)
Visual transition interval pH 8.5 (blue-green)
Visual transition interval pH 9.0 (blue)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR