Diethyl (cyanomethyl)phosphonate 96%

Warning

AC11427-0500 AC11427-2500 AC11427-0100
200018-778EA 54.8 USD
200018-778 200012-030 103524-936
Diethyl (cyanomethyl)phosphonate 96%
Diethyl (cyanomethyl)phosphonate
Formula: C₆H₁₂NO₃P
MW: 177.14 g/mol
Boiling Pt: 101…102 °C (.4 mmHg)
Density: 1.09 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00001893
CAS Number: 2537-48-6
EINECS: 219-806-0
UN: 3278
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4312 to 1.4332 (20 °C, 589 nm)
GC ≥95 %
Additional info Al2O3: 0.040 %
Additional info TiO2: 0.016 %
Additional info K2O: 0.006%
Additional info CaO: 0.005%
Additional info Cr2O3: 0.00005 %
Particle size 40 to 100 mesh
Particle size 205 µm (D50) Typical

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR