Diethyl ethylmalonate 99%

AC11436-1000 AC11436-5000
200043-218EA 78.3 USD
200043-218 200024-996
Diethyl ethylmalonate 99%
Diethyl ethylmalonate
Formula: C₉H₁₆O₄
Boiling Pt: 75…77 °C (5 mmHg)
Density: 1 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00009167
CAS Number: 133-13-1
EINECS: 205-093-3

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4170 (20 °C, 589 nm)
GC ≥98.5 %
Color scale ≤15 APHA
Stabilizer 40 to 60 ppm MEHQ

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR