Diethyl allylmalonate 97%

Warning

AC11396-1000 AC11396-5000
200043-198EA 388 USD
200043-198 200024-988
Diethyl allylmalonate 97%
Diethyl allylmalonate
Formula: C₁₀H₁₆O₄
Density: 1.01 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00009155
CAS Number: 2049-80-1
EINECS: 218-072-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4320 (20 °C, 589 nm)
GC ≥96.0 %
Enantiomeric excess ≥97.5 %
Specific optical rotation +29° to +33° (20 °C, 589 nm) (Neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR