Diethyl acetamidomalonate 99+%

Warning

Synonyms: Diethyl 2-acetamidomalonate

AC11393-1000 AC11393-0250 AC11393-5000
200012-012EA 124 USD
200012-012 200012-010 103509-080
Diethyl acetamidomalonate 99+%
Diethyl acetamidomalonate
Formula: C₉H₁₅NO₅
MW: 217.22 g/mol
Boiling Pt: 185 °C (20 mmHg)
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00009146
CAS Number: 1068-90-2
EINECS: 213-952-9

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 97 °C
GC ≥99.0 %
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR