1,3-Dicyclohexylurea 98%

Synonyms: N,N'-DicyclohexylureaN,N′-Dicyclohexylurea

AC11391-0250 AC11391-1000
200032-064EA 172 USD
200032-064 200019-920
1,3-Dicyclohexylurea 98%
1,3-Dicyclohexylurea
Formula: C₁₃H₂₄N₂O
MW: 224.35 g/mol
Melting Pt: 231…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00003829
CAS Number: 2387-23-7
EINECS: 219-213-7

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 231 to 236 °C
Titration with HClO4 ≥97.5 %
HPLC ≥99.0 % (ELSD)
Dnase,Rnase,Protease None detected
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR