Diallylamine 97%

Danger

AC11204-2500 AC11204-0010
200030-900EA 148 USD
200030-900 200018-722
Diallylamine 97%
Diallylamine
Formula: C₆H₁₁N
Melting Pt: –88 °C
Density: 0.789 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00008642
CAS Number: 124-02-7
EINECS: 204-671-2
UN: 2359
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4395 to 1.4415 (20 °C, 589 nm)
GC ≥96.0 %
UV at 300 nm A: ≤0.50
UV at 320 nm A: ≤0.10
UV at 340 nm A: ≤0.01
UV at 450 nm A: ≤0.01
Color scale ≤20 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR