2-Deoxy-D-glucose 99%

Synonyms: (3R,4S,5R)-3,4,5,6-Tetrahydroxyhexanal

AC11198-0050 AC11198-0010 AC11198-0250
103508-782EA 880 USD
103508-782 200000-286 200009-352
2-Deoxy-D-glucose 99%
2-Deoxy-D-glucose
Formula: C₆H₁₂O₅
MW: 164.16 g/mol
Melting Pt: 146…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00151328
CAS Number: 154-17-6
EINECS: 205-823-0

Order Now

Specification Test Results

Appearance White to cream Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤1 %
Solubility (2 % in water) Clear colorless to light yellow
TLC ≥98.5 %
Residue after ignition ≤1 %
Specific optical rotation +44.0° to +48.0° (20 °C, 589 nm) (c=1, H2O) (after 24 h)
Microbial contamination Passes test
pH 3.0 to 5.0 (2 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR