Cycloheptanecarboxylic acid 97%

Warning

Synonyms: Cycloheptane carboxylic acid

AC11102-0050 AC11102-0250
102775-724EA 109 USD
102775-724 102777-366
Cycloheptanecarboxylic acid 97%
Cycloheptanecarboxylic acid
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 135…138 °C (9 mmHg)
Melting Pt: 71 °C
Density: 1.035 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004152
CAS Number: 1460-16-8

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4710 to 1.4730 (20 °C, 589 nm)
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR