1-(2-Chloroethyl)pyrrolidine hydrochloride 98%

Warning

Synonyms: N-(2-Chloroethyl)pyrrolidine hydrochloride

AC10922-0250 AC10922-1000
200055-912EA 123 USD
200055-912 200038-006
1-(2-Chloroethyl)pyrrolidine hydrochloride 98%
1-(2-Chloroethyl)pyrrolidine hydrochloride

Appearance: White to off-white Crystalline powder or crystals

Formula: C₆H₁₂ClN
Melting Pt: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00012718
CAS Number: 7250-67-1
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 165 to 175 °C
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR