4-Chlorophenethylamine 98%

Danger

Synonyms: 2-(4-Chlorophenyl)ethylamine

AC10988-0050 AC10988-0250
200008-814EA 80.5 USD
200008-814 103530-454
4-Chlorophenethylamine 98%
4-Chlorophenethylamine
Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Boiling Pt: 242…245 °C (760 mmHg)
Density: 1.112 g/cm³
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00008191
CAS Number: 156-41-2
EINECS: 205-853-4
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5474 to 1.5494 (20 °C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR