1-(4-Chlorobenzhydryl)piperazine 90-95%

Danger

Synonyms: 1-(4-Chlorodiphenylmethyl)piperazine1-((4-Chlorophenyl)(phenyl)methyl)piperazine

AC10865-1000 AC10865-0250 AC10865-0050
103508-550EA 621 USD
103508-550 200015-416 200027-482
1-(4-Chlorobenzhydryl)piperazine 90-95%
1-(4-Chlorobenzhydryl)piperazine
Formula: C₁₇H₁₉ClN₂
MW: 286.80 g/mol
Boiling Pt: 178…180 °C (.5 mmHg)
Melting Pt: 65…75 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005965
CAS Number: 303-26-4
UN: 3259

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 65 to 75 °C
Titration Tertiary base ≥90 % (on dry substance)
Water ≤4 % (K.F.)
TLC ≥90 %
Titration with HClO4 ≥90.0 % (On dry substance)
UV at 260 nm A: ≤0.30
UV at 255 nm A: ≤1.0
Color scale ≤10 APHA
Specific gravity (20 °C) 0.900 to 0.904
Residue after evaporation ≤0.0002 %
Acidity ≤0.0008 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR