(Ethoxycarbonylmethyl)triphenylphosphonium bromide ≥98%

Supplier: ACROS ORGANICS MS

Warning

Synonyms: (Carbethoxymethyl)triphenylphosphonium bromide

AC10830-0250
200015-258EA 40.71 USD
200015-258
(Ethoxycarbonylmethyl)triphenylphosphonium bromide ≥98%
(Ethoxycarbonylmethyl)triphenylphosphonium bromide
Formula: C₂₂H₂₂BrO₂P
Storage Temperature: Ambient
MDL Number: MFCD00011835
CAS Number: 1530-45-6

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR