Dibutyl sulfoxide ≥93%, tech.
Synonyms:
n-Butyl sulphoxide
, Butyl sulphoxide
, Di-n-butyl sulphoxide
AC10804-0250
200032-314EA
80.1
USD
200032-314
Dibutyl sulfoxide ≥93%, tech.
Dibutyl sulphoxide
Formula:
C₈H₁₈OS Density: 0,83 g/cm³ Flash Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002093 CAS Number: 2168-93-6 |
Specification Test Results
Appearance | Colorless Low melting crystalline solid |
Infrared spectrum | Conforms |
Melting point | 30°C to 35°C |
GC | ≥91 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...