DL-Carnitine hydrochloride 99%

Danger

Synonyms: 3-Hydroxy-4-(trimethylammonio)butanoate hydrochloride

AC10845-1000 AC10845-0250
200031-126EA 32.8 USD
200031-126 200025-192
DL-Carnitine hydrochloride 99%
DL-Carnitine hydrochloride
Formula: C₇H₁₆ClNO₃
MW: 197.66
Melting Pt: 190…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00011904
CAS Number: 461-05-2
EINECS: 207-309-1

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Melting point 194 to 200 °C
Specific optical rotation -1° to +1° (20 °C, 589 nm) (c=1, H2O)
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR