3-Sulfolene 98%

Danger

Synonyms: Butadiene sulphoneButadiene sulfone

AC10761-0025 AC10761-1000 AC10761-5000
200000-154EA 29.7 USD
200000-154 103524-730 103524-728
3-Sulfolene 98%
3-Sulpholene
Formula: C₄H₆O₂S
MW: 118.16 g/mol
Melting Pt: 63…66 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00005481
CAS Number: 77-79-2
EINECS: 201-059-7
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Adhering powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 63 to 67 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR