L(-)-Bornyl acetate 95%

AC10655-1000 AC10655-5000 AC10655-0025 AC10655-0050
200043-750EA 152 USD
200043-750 200043-752 200031-688 103508-468
L(-)-Bornyl acetate 95%
L(-)-Bornyl acetate
Formula: C₁₂H₂₀O₂
Boiling Pt: 223…224 °C (760 mmHg)
Density: 0.9875 g/cm³
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00867808
CAS Number: 5655-61-8
EINECS: 227-101-4

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4650 (20 °C, 589 nm)
GC ≥94 %
GC ≥98 % (as C12H20O2) Total esters
Specific gravity (15/15 °C) 0.986 to 0.99
Specific optical rotation (20 °C, 589 nm) (neat) < -38°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR