1,2-Di(4-pyridyl)ethane 97%

Warning

Synonyms: 1,2-Bis(4-pyridyl)ethane4,4'-Ethylenedipyridine1,2-Di(pyridin-4-yl)ethane

AC10647-0010
200008-414EA 45.4 USD
200008-414
1,2-Di(4-pyridyl)ethane 97%
1,2-Di(4-pyridyl)ethane
Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Melting Pt: 107…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006451
CAS Number: 4916-57-8

Order Now

Specification Test Results

Appearance White to light yellow to pink Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 107 to 113 °C
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR