Bis(2-chloroethyl)ammonium chloride 98%

Danger

Synonyms: Bis(2-chloroethyl)amine hydrochloride

AC10632-1000
200011-396EA 58.5 USD
200011-396
Bis(2-chloroethyl)ammonium chloride 98%
Bis(2-chloroethyl)ammonium chloride
Formula: C₄H₁₀Cl₃N
MW: 178.49 g/mol
Melting Pt: 210…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00012515
CAS Number: 821-48-7
EINECS: 212-479-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 210 to 215 °C
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR