2,2'-Biquinoline 98%

Warning

AC10630-0010 AC10630-0050
200028-076EA 45.34 USD
200028-076 200008-872
2,2'-Biquinoline 98%
2,2'-Biquinoline
Formula: C₁₈H₁₂N₂
MW: 256.31 g/mol
Melting Pt: 193…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00006740
CAS Number: 119-91-5
EINECS: 204-357-5

Order Now

Specification Test Results

Appearance White to cream to yellow to light brown Powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
Melting point 192 to 198 °C
Titration with HClO4 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR