Benzyltrimethylammonium hydroxide 40% (w/w) in methanol

Danger

Synonyms: Benzyltrimethylammonium HydroxideN,N,N-Trimethyl-1-phenylmethanaminium Hydroxide

AC10620-0051 AC10620-2500 AC10620-0010
200057-170EA 74.5 USD
200057-170 103530-198 200057-164
Benzyltrimethylammonium hydroxide 40% (w/w) in methanol
Benzyltrimethylammonium hydroxide
Formula: C₁₀H₁₇NO
MW: 167.25 g/mol
Density: 0.925 g/cm³
Flash Pt: 15 °C
Storage Temperature: Ambient
EINECS: 202-895-5
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Infrared spectrum Conforms
Titration with HCl ≥40 %
Titration with HClO4 ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR