5-Benzyloxytryptamine hydrochloride 98%

Warning

Synonyms: 2-[5-(Benzyloxy)-1H-indol-3-yl]ethanamine hydrochloride5-(phenylmethoxy)indole-3-ethanamine monohydrochloride

AC10611-0010 AC10611-2500
200042-212EA 139 USD
200042-212 200042-214
5-Benzyloxytryptamine hydrochloride 98%
5-Benzyloxytryptamine hydrochloride
Formula: C₁₇H₁₉ClN₂O
Melting Pt: 255…259 °C
Storage Temperature: Freezer
MDL Number: MFCD00012685
CAS Number: 52055-23-9
EINECS: 257-626-4

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
HPLC ≥97.5 %
Stabilizer 20 to 60 ppm amylene
Residue after evaporation ≤0.002 %
Free halogens passes test
Titrable acid ≤0.0003 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR