Benzoin 98%

Synonyms: 2-Hydroxy-2-phenylacetophenone

AC10551-5000 AC10551-1000 AC10551-0025
103508-410EA 279 USD
103508-410 200005-812 200000-072
Benzoin 98%
Benzoin
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 194 °C (12 mmHg)
Melting Pt: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00004496
CAS Number: 119-53-9
EINECS: 204-331-3

Order Now

Specification Test Results

Appearance Off-white to light yellow Powder
Infrared spectrum Conforms
Melting point 133 to 137 °C
Loss on drying ≤1 %
Sulfated ash ≤0.2 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR