Anthrone ACS

Warning

AC10496-5000
103508-860EA 634.3 USD
103508-860
Anthrone ACS
Anthrone
Formula: C₁₄H₁₀O
MW: 194.23 g/mol
Boiling Pt: 721 °C (760 mmHg)
Melting Pt: 152…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00001187
CAS Number: 90-44-8
EINECS: 201-994-0

Order Now

Specification Test Results

Appearance Yellow Crystals
Infrared spectrum Conforms
Melting point 152 to 157 °C
Solubility (in ethyl acetate) Passes test
Absorbance ≤0.75 (425 nm, 1 cm cell)
Absorbance ≤0.045 (620 nm, 1 cm cell)
Sensitivity (to carbohydrates) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR