4,4'-Anisoin 95%

Synonyms: Anisoin2-Hydroxy-1,2-bis(4-methoxyphenyl)ethanone

AC10484-0250 AC10484-1000
103508-852EA 39.2 USD
103508-852 200024-436
4,4'-Anisoin 95%
4,4'-Anisoin
Formula: C₁₆H₁₆O₄
MW: 272.30 g/mol
Melting Pt: 109…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00008411
CAS Number: 119-52-8
EINECS: 204-330-8

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 108 to 115 °C
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR