5-Aminoquinoline

Warning

Synonyms: Quinolin-5-amineQuinolin-5-ylamine

AC10460-0050 AC10460-0010
102776-542EA 242 USD
102776-542 102774-884
5-Aminoquinoline
5-Aminoquinoline
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 106…109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006797
CAS Number: 611-34-7

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 106 to 112 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR