3-Aminopyrazine-2-carboxylic acid 99+%

Danger

AC10450-0050 AC10450-0250
200023-166EA 162 USD
200023-166 200035-250
3-Aminopyrazine-2-carboxylic acid 99+%
3-Aminopyrazine-2-carboxylic acid
Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Melting Pt: 201…205 °C
Storage Temperature: Ambient
EINECS: 226-558-7

Order Now

Specification Test Results

Appearance Yellow-green to beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥99.0 %
Average molecular weight 16000 (Mn)
Acetyl content ≤4 %
Hydroxyl content 1.5 % (typical)
Butyryl content 50 to 54 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR